Thermo Scientific Chemicals

1,4-Bis(diphenylphosphino)butane, 97%, Thermo Scientific Chemicals

Catalog number: B21122.06
5 g, Each
Thermo Scientific Chemicals

1,4-Bis(diphenylphosphino)butane, 97%, Thermo Scientific Chemicals

Catalog number: B21122.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B21122.06
also known as B21122-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7688-25-7
IUPAC Name
[4-(diphenylphosphanyl)butyl]diphenylphosphane
Molecular Formula
C28H28P2
InChI Key
BCJVBDBJSMFBRW-UHFFFAOYSA-N
SMILES
C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
Proton NMR
Conforms
Form
Powder
Purity
≥96.0%
Appearance (Color)
White

Description

1,4-Bis(diphenylphosphino)butane is used in Suzuki reaction. It plays an essential role as a ligand in palladium(0)-catalyzed acylcyanation of arylacetylenes and deprotection of allyloxycarbamates. It acts as a ligand for alkylations, isomerization reactions and in organometallic chemistry. Further, it is used in Heck reaction, Negishi coupling and Sonogashira coupling. In addition to this, it is useful in organometallic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Bis(diphenylphosphino)butane is used in Suzuki reaction. It plays an essential role as a ligand in palladium(0)-catalyzed acylcyanation of arylacetylenes and deprotection of allyloxycarbamates. It acts as a ligand for alkylations, isomerization reactions and in organometallic chemistry. Further, it is used in Heck reaction, Negishi coupling and Sonogashira coupling. In addition to this, it is useful in organometallic chemistry.

Solubility
Soluble in chloroform. Slightly soluble in water

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text