Thermo Scientific Chemicals

Triisopropylsilyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: B21127.14
25 g, Each
Thermo Scientific Chemicals

Triisopropylsilyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: B21127.14
25 g, Each
Quantity
Catalog number: B21127.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80522-42-5
IUPAC Name
tris(propan-2-yl)silyl trifluoromethanesulfonate
Molecular Formula
C10H21F3O3SSi
InChI Key
LHJCZOXMCGQVDQ-UHFFFAOYSA-N
SMILES
CC(C)[Si](OS(=O)(=O)C(F)(F)F)(C(C)C)C(C)C
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.4120-1.4170 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥96.0%

Description

Triisopropylsilyl trifluoromethanesulfonate is used as a reagent for the introduction of triisopropylsilyl(TIPS) group in organic synthesis. It is involved in the synthesis of 2-substituted benzothiopyran-4-ones and silyloxy acetylenes. As a protecting group in organic synthesis, it is utilized especially for the protection of primary amines. Furthermore, it plays an important role in Takahashi Taxol total synthesis or for chemical glycosylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triisopropylsilyl trifluoromethanesulfonate is used as a reagent for the introduction of triisopropylsilyl(TIPS) group in organic synthesis. It is involved in the synthesis of 2-substituted benzothiopyran-4-ones and silyloxy acetylenes. As a protecting group in organic synthesis, it is utilized especially for the protection of primary amines. Furthermore, it plays an important role in Takahashi Taxol total synthesis or for chemical glycosylation reactions.

Solubility
Miscible with chloroform and ethyl acetate.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents. Hydrolyzes in water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text