3,4,5-Trimethoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
3,4,5-Trimethoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4,5-Trimethoxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B21136.22
also known as B21136-22
Price (USD)/ Each
631.65
Online exclusive
702.00 
Save 70.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
631.65
Online exclusive
702.00 
Save 70.35 (10%)
Add to cart
3,4,5-Trimethoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
Catalog numberB21136.22
Price (USD)/ Each
631.65
Online exclusive
702.00 
Save 70.35 (10%)
-
Add to cart
Chemical Identifiers
CAS951-82-6
IUPAC Name2-(3,4,5-trimethoxyphenyl)acetic acid
Molecular FormulaC11H14O5
InChI KeyDDSJXCGGOXKGSJ-UHFFFAOYSA-N
SMILESCOC1=CC(CC(O)=O)=CC(OC)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Melting Point (clear melt)116.5-122.5?C
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
View more
It is used in the synthesis of 3,4-Diaryl-2(5H)-furanones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of 3,4-Diaryl-2(5H)-furanones.

Solubility
Soluble in water. Solubility in methanol is almost transparent.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Elliot Spector. Identification of 3,4,5-Trimethoxyphenylacetic Acid as the Major Metabolite of Mescaline in the Dog.Nature.1961 751-752.
  2. Yong Kim, et al. Synthesis and Cytotoxicity of 3,4-Diaryl-2(5H)-furanones.Bioorg. Med. Chem. Lett.200212(4), 719-722.