Thermo Scientific Chemicals

Isopropyl-beta-D-thiogalactoside, dioxane-free, 99%, Thermo Scientific Chemicals

Catalog number: B21149.06
5g, Each
Thermo Scientific Chemicals

Isopropyl-beta-D-thiogalactoside, dioxane-free, 99%, Thermo Scientific Chemicals

Catalog number: B21149.06
5g, Each
Quantity
Catalog number: B21149.06
also known as B21149-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
367-93-1
IUPAC Name
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
Molecular Formula
C9H18O5S
InChI Key
BPHPUYQFMNQIOC-NXRLNHOXSA-N
SMILES
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Appearance (Color)
White to pale cream
Form
Powder or crystalline powder
Assay (Silylated GC)
≥98.5% (non-U.S. specification)
Assay from Suppliers CofA
≥98.0% (HPLC: U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

Beta-galactosidase inducer.Isopropyl-β-D-thiogalactoside is used as a reagent in molecular biology. It is used as an effective inducer of protein expression where the gene is under the control of the lac operator. It is used with 5-bromo-4-chloro-3-indolyl-beta-D-galactopyranoside to identify the non-recombinant plasmid in cloning experiments by blue-white screen methodology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Beta-galactosidase inducer.Isopropyl-β-D-thiogalactoside is used as a reagent in molecular biology. It is used as an effective inducer of protein expression where the gene is under the control of the lac operator. It is used with 5-bromo-4-chloro-3-indolyl-beta-D-galactopyranoside to identify the non-recombinant plasmid in cloning experiments by blue-white screen methodology.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text