Thermo Scientific Chemicals

Diethyl 2-bromoethylphosphonate, 97%, Thermo Scientific Chemicals

Catalog number: B21156.14
25 g, Each
Thermo Scientific Chemicals

Diethyl 2-bromoethylphosphonate, 97%, Thermo Scientific Chemicals

Catalog number: B21156.14
25 g, Each
Quantity
Catalog number: B21156.14
also known as B21156-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5324-30-1
IUPAC Name
diethyl (2-bromoethyl)phosphonate
Molecular Formula
C6H14BrO3P
InChI Key
PINITSMLVXAASM-UHFFFAOYSA-N
SMILES
CCOP(=O)(CCBr)OCC
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Refractive Index
1.4585-1.4635 @ 20?C
Form
Liquid

Description

It is employed as a reactant involved in radical coupling, synthesis of organosoluble zirconium phosphonate nanocomposites, click-chemistry, asymmetric epoxidation of unfunctionalized olefins, Negishi alkyl-aryl cross-coupling, and in synthesis of hydrolytically stable phosphonic acids for use as dental adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reactant involved in radical coupling, synthesis of organosoluble zirconium phosphonate nanocomposites, click-chemistry, asymmetric epoxidation of unfunctionalized olefins, Negishi alkyl-aryl cross-coupling, and in synthesis of hydrolytically stable phosphonic acids for use as dental adhesives.

Solubility
Soluble in water(20 g/L) (25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text