Thermo Scientific Chemicals

Dimethyl adipate, 99%, Thermo Scientific Chemicals

Catalog number: B21174.18
50 g, Each
Thermo Scientific Chemicals

Dimethyl adipate, 99%, Thermo Scientific Chemicals

Catalog number: B21174.18
50 g, Each
Quantity
Catalog number: B21174.18
also known as B21174-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
627-93-0
IUPAC Name
1,6-dimethyl hexanedioate
Molecular Formula
C8H14O4
InChI Key
UDSFAEKRVUSQDD-UHFFFAOYSA-N
SMILES
COC(=O)CCCCC(=O)OC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.4270-1.4300 @ 20?C

Description

Dimethyl adipate is used in cosmetics (in emollients and skin conditioning). It acts as a cosmetic plasticizer. It is also used in agrochemicals and dye as well as a precursor for the preparation of active pharmaceutical ingredients. Further, it serves as a polymer intermediate. In addition to this, it is employed as a solvent for paint stripping and resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl adipate is used in cosmetics (in emollients and skin conditioning). It acts as a cosmetic plasticizer. It is also used in agrochemicals and dye as well as a precursor for the preparation of active pharmaceutical ingredients. Further, it serves as a polymer intermediate. In addition to this, it is employed as a solvent for paint stripping and resins.

Solubility
Miscible with alcohols and ether. Immiscible with water.

Notes
Moisture sensitive. Incompatible with acids, bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text