Trimethoxyvinylsilane is used as a chemical intermediate in the production of other fine chemicals or as a monomer in the production of vinyl functional silicone polymers. The use of Trimethoxyvinylsilane in coatings is intended to improve bonding or adhesion between organic polymers and mineral surfaces, such as pigments, fillers, and glass or metal substrates. It is used in water-based decorative coating products. It is also used as a as cross-linking agent and adhesion promoter in commercially-available silicone sealant product.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trimethoxyvinylsilane is used as a chemical intermediate in the production of other fine chemicals or as a monomer in the production of vinyl functional silicone polymers. The use of Trimethoxyvinylsilane in coatings is intended to improve bonding or adhesion between organic polymers and mineral surfaces, such as pigments, fillers, and glass or metal substrates. It is used in water-based decorative coating products. It is also used as a as cross-linking agent and adhesion promoter in commercially-available silicone sealant product.
Solubility
Solubility in water, 9.4 g/L at 20°C.
Notes
Moisture sensitive. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only
General References:
- Hyun-Min Kim, et al. Biomimetic apatite formation on polyethylene photografted with vinyltrimethoxysilane and hydrolyzed.Biomater.200122(18), 2489-2494.
- B. Naderi Zand, et al. Evaluation of the effect of vinyltrimethoxysilane on corrosion resistance and adhesion strength of epoxy coated AA1050.Electrochim. Acta,200752(23), 6438-6442.