D-Threonine, 99%
Thermo Scientific Chemicals

D-Threonine, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B21177.06
also known as B21177-06
Price (USD)/ Each
40.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
40.30
Add to cart
D-Threonine, 99%
Catalog numberB21177.06
Price (USD)/ Each
40.30
-
Add to cart
Chemical Identifiers
CAS632-20-2
IUPAC Name(2R,3S)-2-amino-3-hydroxybutanoic acid
Molecular FormulaC4H9NO3
InChI KeyAYFVYJQAPQTCCC-STHAYSLISA-N
SMILESC[C@H](O)[C@@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Optical Rotation+29? ? 2? (c=5, water)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only