Thermo Scientific Chemicals

4-Nitrophenylacetone, 98%, Thermo Scientific Chemicals

Catalog number: B21183.03
1 g, Each
Thermo Scientific Chemicals

4-Nitrophenylacetone, 98%, Thermo Scientific Chemicals

Catalog number: B21183.03
1 g, Each
Quantity
Catalog number: B21183.03
also known as B21183-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5332-96-7
IUPAC Name
1-(4-nitrophenyl)propan-2-one
Molecular Formula
C9H9NO3
InChI Key
GEWWCWZGHNIUBW-UHFFFAOYSA-N
SMILES
CC(=O)CC1=CC=C(C=C1)[N+]([O-])=O
Appearance (Color)
Yellow to orange
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
60.0-67.0?C

Description

It is applied as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an intermediate in organic synthesis.

Solubility
Insoluble in water, soluble in usual organic solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text