Thermo Scientific Chemicals

(3-Mercaptopropyl)triethoxysilane, 94%, Thermo Scientific Chemicals

Catalog number: B21191.22
100 g, Each
Thermo Scientific Chemicals

(3-Mercaptopropyl)triethoxysilane, 94%, Thermo Scientific Chemicals

Catalog number: B21191.22
100 g, Each
Quantity
Catalog number: B21191.22
also known as B21191-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14814-09-6
IUPAC Name
3-(triethoxysilyl)propane-1-thiol
Molecular Formula
C9H22O3SSi
InChI Key
DCQBZYNUSLHVJC-UHFFFAOYSA-N
SMILES
CCO[Si](CCCS)(OCC)OCC
Identification (FTIR)
Conforms
Refractive Index
1.4340-1.4410 @ 20?C
Appearance (Color)
Colorless
Form
Crystals or powder or crystalline powder or clear liquid as melt
Assay (GC)
≥92.0%

Description

It can improve the properties of the mineral-filled elastomer, including modulus, tensile and tear strength, heat buildup, abrasion resistance, resilience. It can be used as a pretreatment on minerals or added at the time of compounding. Mineral-reinforced rubbers such as shoe soles, rollers, wheels, white sidewalls, wire and cable insulation are the common case (3-Mercaptopropyl)triethoxysilane, could be used in. It also finds it uses as an intermediate, in polymer and tire manufacture, and as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can improve the properties of the mineral-filled elastomer, including modulus, tensile and tear strength, heat buildup, abrasion resistance, resilience. It can be used as a pretreatment on minerals or added at the time of compounding. Mineral-reinforced rubbers such as shoe soles, rollers, wheels, white sidewalls, wire and cable insulation are the common case (3-Mercaptopropyl)triethoxysilane, could be used in. It also finds it uses as an intermediate, in polymer and tire manufacture, and as a laboratory reagent.

Solubility
Hydrolyzes slowly in water.

Notes
Air & moisture sensitive. Store away from oxidizing agents, air and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text