Thermo Scientific Chemicals

cis,cis-1,5-Cyclooctadiene, 99%, stab. with 50-200ppm Irganox 1076, Thermo Scientific Chemicals

Catalog number: B21196.AK
250 mL, Each
Thermo Scientific Chemicals

cis,cis-1,5-Cyclooctadiene, 99%, stab. with 50-200ppm Irganox 1076, Thermo Scientific Chemicals

Catalog number: B21196.AK
250 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B21196.AK
also known as B21196-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-78-4
IUPAC Name
(1Z,5Z)-cycloocta-1,5-diene
Molecular Formula
C8H12
InChI Key
VYXHVRARDIDEHS-QGTKBVGQSA-N
SMILES
C1C\C=C/CC\C=C/1
Appearance (Color)
Clear, colourless
Form
liquid
Assay (GC)
> 98.5%
Refractive Index
1.4925 - 1.4955 @20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text