D-Sorbose, 98%
Thermo Scientific Chemicals

D-Sorbose, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.1 g
0.5 g
Catalog number B21208.77
also known as B21208-77
Price (USD)/ Each
95.00
-
Add to cart
Quantity:
0.1 g
Request bulk or custom format
Price (USD)/ Each
95.00
Add to cart
D-Sorbose, 98%
Catalog numberB21208.77
Price (USD)/ Each
95.00
-
Add to cart
Chemical Identifiers
CAS3615-56-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (HPLC)≥97.5%
Melting Point (clear melt)158.0-165.0?C
Optical Rotation38.7 ± 0.1? (C=5 in water)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only