Thermo Scientific Chemicals

1-Naphthaleneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B21219.14
25 g, Each
Thermo Scientific Chemicals

1-Naphthaleneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B21219.14
25 g, Each
Quantity
Catalog number: B21219.14
also known as B21219-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13922-41-3
IUPAC Name
(naphthalen-1-yl)boronic acid
Molecular Formula
C10H9BO2
InChI Key
HUMMCEUVDBVXTQ-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CC2=CC=CC=C12
Proton NMR
Conforms to structure
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0%
Assay (HPLC)
≥96.0%

Description

1-Naphthaleneboronic acid is a very useful building block for introduction of a naphthyl group through cross-coupling protocols. It is useful in the synthesis of polyaromatic hydrocarbons utilizing the Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthaleneboronic acid is a very useful building block for introduction of a naphthyl group through cross-coupling protocols. It is useful in the synthesis of polyaromatic hydrocarbons utilizing the Suzuki reaction.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in a well sealed container. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text