Thermo Scientific Chemicals

4-Heptanone, 98%, Thermo Scientific Chemicals

Catalog number: B21222.AE
100 mL, Each
Thermo Scientific Chemicals

4-Heptanone, 98%, Thermo Scientific Chemicals

Catalog number: B21222.AE
100 mL, Each
Quantity
Catalog number: B21222.AE
also known as B21222-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-19-3
IUPAC Name
heptan-4-one
Molecular Formula
C7H14O
InChI Key
HCFAJYNVAYBARA-UHFFFAOYSA-N
SMILES
CCCC(=O)CCC
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Refractive Index
1.4045-1.4085 @ 20?C
Form
Liquid

Description

4-Heptanone is used as a solvent for nitrocellulose as well as in organic synthesis. It is involved in the preparation of flavorings. It is used as an intermediate in the synthesis of active pharmaceutical ingredients. Further, it is used in paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Heptanone is used as a solvent for nitrocellulose as well as in organic synthesis. It is involved in the preparation of flavorings. It is used as an intermediate in the synthesis of active pharmaceutical ingredients. Further, it is used in paints and coatings.

Solubility
Miscible with alcohol, ligroin and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text