Thermo Scientific Chemicals

D-Turanose, 98%

Catalog number: B21224.06
5 g, Each
Thermo Scientific Chemicals

D-Turanose, 98%

Catalog number: B21224.06
5 g, Each
Quantity
Catalog number: B21224.06
also known as B21224-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
547-25-1
Appearance (Color)
White
Optical Rotation
76.0 ± 1.5? (C=4 in water)
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms
Form
Powder

Description

D-Turanose is used as a carbon source by bacteria, fungi, and other organisms. It is involved in intracellular sugar signaling in plant cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Turanose is used as a carbon source by bacteria, fungi, and other organisms. It is involved in intracellular sugar signaling in plant cells.

Solubility
Soluble in water and methanol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in a well sealed container. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text