Thermo Scientific Chemicals

alpha-D-(+)-Talose, 97%, Thermo Scientific Chemicals

Catalog number: B21226.02
0.5 g, Each
Thermo Scientific Chemicals

alpha-D-(+)-Talose, 97%, Thermo Scientific Chemicals

Catalog number: B21226.02
0.5 g, Each
Quantity
Catalog number: B21226.02
also known as B21226-02
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2595-98-4
IUPAC Name
2,3,4,5,6-pentahydroxyhexanal
Molecular Formula
C6H12O6
InChI Key
GZCGUPFRVQAUEE-UHFFFAOYNA-N
SMILES
OCC(O)C(O)C(O)C(O)C=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥96.0%
Melting Point (clear melt)
125.0-131.0?C
Optical Rotation
21? ± 1.0? (c=1 in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text