Thermo Scientific Chemicals

2-Phenylethyl acetate, 98%, Thermo Scientific Chemicals

Catalog number: B21238.0B
1000 g, Each
Thermo Scientific Chemicals

2-Phenylethyl acetate, 98%, Thermo Scientific Chemicals

Catalog number: B21238.0B
1000 g, Each
Quantity
Catalog number: B21238.0B
also known as B21238-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-45-7
IUPAC Name
2-phenylethyl acetate
Molecular Formula
C10H12O2
InChI Key
MDHYEMXUFSJLGV-UHFFFAOYSA-N
SMILES
CC(=O)OCCC1=CC=CC=C1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4960-1.5000 @ 20?C

Description

Phenylethyl acetate mantains the typical rose notes of Phenylethanol with a more intense honey profile which is perfect for fruit and alcoholic drink applications. It is used as a pharmaceutical intermediate and also it holds application in gas chromatography and liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylethyl acetate mantains the typical rose notes of Phenylethanol with a more intense honey profile which is perfect for fruit and alcoholic drink applications. It is used as a pharmaceutical intermediate and also it holds application in gas chromatography and liquid chromatography.

Solubility
It is insoluble in paraffin oil, water. Soluble in alcohol, dipropylene glycol, fixed oils, kerosene, propylene glycol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Protected from heat and light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text