Methyl 2-octynoate, 98%, Thermo Scientific Chemicals
Methyl 2-octynoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 2-octynoate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B21242.14
also known as B21242-14
Price (USD)/ Each
38.90
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
38.90
Add to cart
Methyl 2-octynoate, 98%, Thermo Scientific Chemicals
Catalog numberB21242.14
Price (USD)/ Each
38.90
-
Add to cart
Chemical Identifiers
CAS111-12-6
IUPAC Namemethyl oct-2-ynoate
Molecular FormulaC9H14O2
InChI KeyFRLZQXRXIKQFNS-UHFFFAOYSA-N
SMILESCCCCCC#CC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4455-1.4495 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only