Thermo Scientific Chemicals

(R)-(+)-Styrene oxide, 98%, Thermo Scientific Chemicals

Catalog number: B21245.14
25 g, Each
Thermo Scientific Chemicals

(R)-(+)-Styrene oxide, 98%, Thermo Scientific Chemicals

Catalog number: B21245.14
25 g, Each
Quantity
Catalog number: B21245.14
also known as B21245-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20780-53-4
IUPAC Name
(2R)-2-phenyloxirane
Molecular Formula
C8H8O
InChI Key
AWMVMTVKBNGEAK-QMMMGPOBSA-N
SMILES
C1O[C@@H]1C1=CC=CC=C1
Refractive Index
1.5320-1.5360 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Optical Rotation
33.8 ± 0.1? (neat)

Description

It is used to produce a homologated epoxide as part of a synthetic approach to (+)-allosedamine. It is the main metabolite of styrene, catalyzed by epoxide hydrolase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to produce a homologated epoxide as part of a synthetic approach to (+)-allosedamine. It is the main metabolite of styrene, catalyzed by epoxide hydrolase.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text