1,12-Dibromododecane, is used as an organic chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,12-Dibromododecane, is used as an organic chemical intermediate.
Solubility
Soluble in hot methanol- almost transparency.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Stable. Combustible. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- R Zana.; M Benrraou.; R Rueff. Alkanediyl-.alpha.,.μ.-bis(dimethylalkylammonium bromide) surfactants. 1. Effect of the spacer chain length on the critical micelle concentration and micelle ionization degree. Langmuir. 19917 (6), 1072-1075.