Thermo Scientific Chemicals

1,12-Dibromododecane, 98%, Thermo Scientific Chemicals

Catalog number: B21254.14
25 g, Each
Thermo Scientific Chemicals

1,12-Dibromododecane, 98%, Thermo Scientific Chemicals

Catalog number: B21254.14
25 g, Each
Quantity
Catalog number: B21254.14
also known as B21254-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3344-70-5
IUPAC Name
1,12-dibromododecane
Molecular Formula
C12H24Br2
InChI Key
ZJJATABWMGVVRZ-UHFFFAOYSA-N
SMILES
BrCCCCCCCCCCCCBr
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
36.0-43.0?C

Description

1,12-Dibromododecane, is used as an organic chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,12-Dibromododecane, is used as an organic chemical intermediate.

Solubility
Soluble in hot methanol- almost transparency.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Stable. Combustible. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text