Diphenylmethane, 99+%, Thermo Scientific Chemicals
Diphenylmethane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenylmethane, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number B21256.0E
also known as B21256-0E
Price (USD)/ Each
191.00
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
191.00
Add to cart
Diphenylmethane, 99+%, Thermo Scientific Chemicals
Catalog numberB21256.0E
Price (USD)/ Each
191.00
-
Add to cart
Chemical Identifiers
CAS101-81-5
IUPAC Namebenzylbenzene
Molecular FormulaC13H12
InChI KeyCZZYITDELCSZES-UHFFFAOYSA-N
SMILESC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid or low melting solid
Assay (GC)≥99.0%
Identification (FTIR)Conforms
Refractive Index1.5765-1.5790 @ 20°C
Diphenylmethane, is used as an adhesive chemical composition for making flexible laminates for use in food packaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylmethane, is used as an adhesive chemical composition for making flexible laminates for use in food packaging.

Solubility
Soluble in ethanol, ether, benzene and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. WP Yang.; CW Macosko.; ST Wellinghoff. Thermal degradation of urethanes based on 4, 4'-diphenylmethane diisocyanate and 1, 4-butanediol (MDI/BDO). Polymer. 198627 (8), 1235-1240.