Thermo Scientific Chemicals

4-tert-Butylbenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: B21260.06
5 g, Each
Thermo Scientific Chemicals

4-tert-Butylbenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: B21260.06
5 g, Each
Quantity
Catalog number: B21260.06
also known as B21260-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
877-65-6
IUPAC Name
(4-tert-butylphenyl)methanol
Molecular Formula
C11H16O
InChI Key
FVEINXLJOJPHLH-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=C(CO)C=C1
Assay (GC)
≥97.5%
Refractive Index
1.5160-1.5200 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid

Description

4-tert-Butylbenzyl alcohol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylbenzyl alcohol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in alcohol and insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Keep away from strong oxidizing agents, acid chlorides, acid anhydrides, strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text