Thermo Scientific Chemicals

Serotonin hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: B21263.09
10 g, Each
Thermo Scientific Chemicals

Serotonin hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: B21263.09
10 g, Each
Quantity
Catalog number: B21263.09
also known as B21263-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
153-98-0
IUPAC Name
hydrogen 3-(2-aminoethyl)-1H-indol-5-ol chloride
Molecular Formula
C10H13ClN2O
InChI Key
MDIGAZPGKJFIAH-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCCC1=CNC2=CC=C(O)C=C12
Appearance (Color)
White to pale cream or pale grey or pale brown
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥97.5 to ≤102.5% (Dry wt. basis, material sourced in UK)
Assay from Supplier's CofA
≥97.5% (HPLC)
Comment
Material sourced in UK or US

Description

Serotonin hydrochloride is used as a neurotransmitter and function as an endogenous serotonin receptor agonist in various biochemical processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Serotonin Hydrochloride is used as a neurotransmitter and function as an endogenous serotonin receptor agonist in various biochemical processes.

Solubility
Soluble in water, alcohol, hydrochloric acid and dimethyl sulfoxide. Insoluble in ether and chloroform.

Notes
Store in a cool place. Light sensitive and hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text