3-n-Propylphenol, is used to produce 2,4,5,6-tetrabromo-3-propylphenol. This reaction will need reagent bromine, aluminum powder.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-n-Propylphenol, is used to produce 2,4,5,6-tetrabromo-3-propylphenol. This reaction will need reagent bromine, aluminum powder.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Curt M. White.; Norman C. Li. Determination of phenols in a coal liquefaction product by gas chromatography and combined gas chromatography-mass spectrometry. Anal. Chem. 1982, 54, (9), 1570-1572.
Istvan Ujvary.; Gyula Mikite. A Practical Synthesis of 3-n-Propylphenol, a Component of Tsetse Fly Attractant Blends. Org. Proc. Res. Dev. 2003, 7, (4), 585-587.