2-Bromo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals
2-Bromo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
50 g
Catalog number B21272.09
also known as B21272-09
Price (USD)/ Each
58.10
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
58.10
Add to cart
2-Bromo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals
Catalog numberB21272.09
Price (USD)/ Each
58.10
-
Add to cart
Chemical Identifiers
CAS41825-73-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to brown
FormCrystals or powder or crystalline powder or pellets
Assay (GC)≥97.5%
Water Content (Karl Fischer Titration)≤1%
Melting Point (clear melt)41.0-47.0?C
2-Bromo-4,6-dimethylaniline can react with 3-chloro-3-methyl-but-1-yne to produce 2-bromo-N-(1',1'-dimethylprop-2'-ynyl)-4,6-dimethylaniline. This reaction will need reagent Et3N, catalyst CuCl, and the menstruum dioxane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4,6-dimethylaniline can react with 3-chloro-3-methyl-but-1-yne to produce 2-bromo-N-(1′,1′-dimethylprop-2′-ynyl)-4,6-dimethylaniline. This reaction will need reagent Et3N, catalyst CuCl, and the menstruum dioxane.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Rattan K. Gujadhur.; Craig G. Bates.; D. Venkataraman. Formation of Aryl-Nitrogen, Aryl-Oxygen, and Aryl-Carbon Bonds Using Well-Defined Copper(I)-Based Catalysts. Org. Lett. 2001, 3, (26), 4315-4317.
  2. Fuk Yee Kwong.; Artis Klapars.; Stephen L. Buchwald. Copper-Catalyzed Coupling of Alkylamines and Aryl Iodides: An Efficient System Even in an Air Atmospheres. Org. Lett. 2002, 4, (4), 581-584.