Thermo Scientific Chemicals

2-Bromo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals

Catalog number: B21272.14
25 g, Each
Thermo Scientific Chemicals

2-Bromo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals

Catalog number: B21272.14
25 g, Each
Quantity
Catalog number: B21272.14
also known as B21272-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
41825-73-4
Appearance (Color)
Cream to brown
Form
Crystals or powder or crystalline powder or pellets
Assay (GC)
≥97.5%
Water Content (Karl Fischer Titration)
≤1%
Melting Point (clear melt)
41.0-47.0?C

Description

2-Bromo-4,6-dimethylaniline can react with 3-chloro-3-methyl-but-1-yne to produce 2-bromo-N-(1',1'-dimethylprop-2'-ynyl)-4,6-dimethylaniline. This reaction will need reagent Et3N, catalyst CuCl, and the menstruum dioxane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4,6-dimethylaniline can react with 3-chloro-3-methyl-but-1-yne to produce 2-bromo-N-(1′,1′-dimethylprop-2′-ynyl)-4,6-dimethylaniline. This reaction will need reagent Et3N, catalyst CuCl, and the menstruum dioxane.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text