2-Cyano-4'-methylbiphenyl, 98+%, Thermo Scientific Chemicals
2-Cyano-4'-methylbiphenyl, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Cyano-4'-methylbiphenyl, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B21273.14
also known as B21273-14
Price (USD)/ Each
137.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
162.00 
Save 24.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
137.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
162.00 
Save 24.35 (15%)
Add to cart
2-Cyano-4'-methylbiphenyl, 98+%, Thermo Scientific Chemicals
Catalog numberB21273.14
Price (USD)/ Each
137.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
162.00 
Save 24.35 (15%)
-
Add to cart
Chemical Identifiers
CAS114772-53-1
IUPAC Name4'-methyl-[1,1'-biphenyl]-2-carbonitrile
Molecular FormulaC14H11N
InChI KeyZGQVZLSNEBEHFN-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder or flakes
Assay (GC)≥98.0%
Melting Point (clear melt)48.0-54.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only