3-Formylbenzoic acid is used in the synthesis of bicyclic cis-2-azetidinone derivatives via Ugi 4-centre 3-component reaction, porphyrin capped with a steroidal superstructure bearing convergent hydroxy groups and 3-[(4-amino-1,2-dihydro-1-oxo-2-phenyl-1,2,4-triazolo[4,3-a]quinoxalin-6-yl)amino]methyl benzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Formylbenzoic acid is used in the synthesis of bicyclic cis-2-azetidinone derivatives via Ugi 4-centre 3-component reaction, porphyrin capped with a steroidal superstructure bearing convergent hydroxy groups and 3-[(4-amino-1,2-dihydro-1-oxo-2-phenyl-1,2,4-triazolo[4,3-a]quinoxalin-6-yl)amino]methyl benzoic acid.
Solubility
Soluble in methanol (100 mg/mL) gives a clear to slightly hazy, colorless to very faintly brown.
Notes
Air sensitive. Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Gedey S, et al. Synthesis of Alicyclic-lactams via the Ugi Reaction on a Solid Support.Lett. Org. Chem.20041(3), 215-20.
Richard P and Jeremy KM. Synthesis, binding properties and self-functionalization of a steroid-capped porphyrin.J. Chem. Soc. Chem. Commun.19918574-77.