Thermo Scientific Chemicals

Dibenzyl diselenide, 95%, Thermo Scientific Chemicals

Catalog number: B21278.14
25 g, Each
Thermo Scientific Chemicals

Dibenzyl diselenide, 95%, Thermo Scientific Chemicals

Catalog number: B21278.14
25 g, Each
Quantity
Catalog number: B21278.14
also known as B21278-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1482-82-2
IUPAC Name
[(benzyldiselanyl)methyl]benzene
Molecular Formula
C14H14Se2
InChI Key
HYAVEDMFTNAZQE-UHFFFAOYSA-N
SMILES
C([Se][Se]CC1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Yellow to brown
Form
Powder
Assay (unspecified)
≥94.0%
Comment
Specification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)
Conforms (non-U.S. specification)

Description

Dibenzyl diselenide is used as a source of the PhSe unit in organic synthesis. It is also used to introduce PhSe groups by reaction with a variety of nucleophilic, including enolates, enol silyl ethers, Grignard reagents, organolithium reagents, alkenes and amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl diselenide is used as a source of the PhSe unit in organic synthesis. It is also used to introduce PhSe groups by reaction with a variety of nucleophilic, including enolates, enol silyl ethers, Grignard reagents, organolithium reagents, alkenes and amines.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text