Thermo Scientific Chemicals

Isoquinoline, 97%, Thermo Scientific Chemicals

Catalog number: B21279.22
100 g, Each
Thermo Scientific Chemicals

Isoquinoline, 97%, Thermo Scientific Chemicals

Catalog number: B21279.22
100 g, Each
Quantity
Catalog number: B21279.22
also known as B21279-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
119-65-3
IUPAC Name
isoquinoline
Molecular Formula
C9H7N
InChI Key
AWJUIBRHMBBTKR-UHFFFAOYSA-N
SMILES
C1=CC=C2C=NC=CC2=C1
Appearance (Color)
Cream to yellow to brown to purple
Form
Crystals or powder or crystalline powder or clear liquid as melt
Assay (GC)
≥97.0%
Identification (FTIR)
Conforms
Refractive Index
1.6215-1.6265 @ 20?C

Description

Isoquinoline is used in the manufacture of dyes, paints, antifungal. It is also used as a solvent for the extraction of resins and terpenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isoquinoline is used in the manufacture of dyes, paints, antifungal. It is also used as a solvent for the extraction of resins and terpenes.

Solubility
Soluble in water (5g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text