Thermo Scientific Chemicals

6-Thioguanine, 98%, Thermo Scientific Chemicals

Catalog number: B21280.03
1 g, Each
Thermo Scientific Chemicals

6-Thioguanine, 98%, Thermo Scientific Chemicals

Catalog number: B21280.03
1 g, Each
Quantity
Catalog number: B21280.03
also known as B21280-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
154-42-7
IUPAC Name
2-amino-6,7-dihydro-3H-purine-6-thione
Molecular Formula
C5H5N5S
InChI Key
WYWHKKSPHMUBEB-UHFFFAOYSA-N
SMILES
NC1=NC(=S)C2=C(N1)N=CN2
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (HPLC)
≥97.5%
Appearance (Color)
White to pale cream or pale yellow or pale yellow-green
Form
Powder

Description

6-Thioguanine acts as an antineoplastic and purine antimetabolite. It is also useful as an inhibition of stimulated expression of TNF-related apoptosis-inducing ligand (TRAIL) protein. It is involved in the treatment of acute leukemias and Psoriasis. Further, it is used for the treatment of ulcerative colitis and autoimmune diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dilute solutions of alkali hydroxides and hot alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text