Thermo Scientific Chemicals

1,4-Dibromo-2-fluorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: B21281.22
100 g, Each
Thermo Scientific Chemicals

1,4-Dibromo-2-fluorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: B21281.22
100 g, Each
Quantity
Catalog number: B21281.22
also known as B21281-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1435-52-5
IUPAC Name
1,4-dibromo-2-fluorobenzene
Molecular Formula
C6H3Br2F
InChI Key
WNSNPGHNIJOOPM-UHFFFAOYSA-N
SMILES
FC1=CC(Br)=CC=C1Br
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥97.5%
Melting Point (clear melt)
29.0-36.0?C

Description

It is used in preparation of 1,4-bis(2-hydroxy-2-methyl-3-butynyl)-2-fluorobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in preparation of 1,4-bis(2-hydroxy-2-methyl-3-butynyl)-2-fluorobenzene.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text