N-(Hydroxymethyl)phthalimide, 97%, Thermo Scientific Chemicals
N-(Hydroxymethyl)phthalimide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(Hydroxymethyl)phthalimide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B21292.14
also known as B21292-14
Price (USD)/ Each
28.80
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
28.80
Add to cart
N-(Hydroxymethyl)phthalimide, 97%, Thermo Scientific Chemicals
Catalog numberB21292.14
Price (USD)/ Each
28.80
-
Add to cart
Chemical Identifiers
CAS118-29-6
IUPAC Name2-(hydroxymethyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC9H7NO3
InChI KeyMNSGOOCAMMSKGI-UHFFFAOYSA-N
SMILESOCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)138-148?C
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only