Thermo Scientific Chemicals

Diisopropyl sulfide, 99%, Thermo Scientific Chemicals

Catalog number: B21302.09
10 g, Each
Thermo Scientific Chemicals

Diisopropyl sulfide, 99%, Thermo Scientific Chemicals

Catalog number: B21302.09
10 g, Each
Quantity
Catalog number: B21302.09
also known as B21302-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
625-80-9
IUPAC Name
2-(propan-2-ylsulfanyl)propane
Molecular Formula
C6H14S
InChI Key
XYWDPYKBIRQXQS-UHFFFAOYSA-N
SMILES
CC(C)SC(C)C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.4370-1.4400 @ 20?C

Description

Used as a flavoring agents and intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavoring agents and intermediate for organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text