Thermo Scientific Chemicals

CAPSO, 98%

Catalog number: B21305.22
100 g, Each
Thermo Scientific Chemicals

CAPSO, 98%

Catalog number: B21305.22
100 g, Each
Quantity
Catalog number: B21305.22
also known as B21305-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
73463-39-5
IUPAC Name
(2S)-3-(cyclohexylazaniumyl)-2-hydroxypropane-1-sulfonate
Molecular Formula
C9H19NO4S
InChI Key
INEWUCPYEUEQTN-VIFPVBQESA-N
SMILES
O[C@@H](C[NH2+]C1CCCCC1)CS([O-])(=O)=O
Appearance (Color)
White
Form
Crystalline powder or flake
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
263-273?C

Description

CAPSO is used in various biological applications such as electrophoresis. Specific applications include buffers for protein separations and diagnostic reagent production. Also used as biological buffer in the following studies, to maintain pH 9 during water leach test for the analysis of high-carbon coal fly ash-soil mixtures, chemiluminescent in situ hybridization using soybean peroxidase-labeled peptide nucleic acid (PNA) probes, preparation of planar bilayer lipid membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
CAPSO is used in various biological applications such as electrophoresis. Specific applications include buffers for protein separations and diagnostic reagent production. Also used as biological buffer in the following studies, to maintain pH 9 during water leach test for the analysis of high-carbon coal fly ash-soil mixtures, chemiluminescent in situ hybridization using soybean peroxidase-labeled peptide nucleic acid (PNA) probes, preparation of planar bilayer lipid membranes.

Solubility
Soluble in water (10 g+ 90 ml).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text