N-Vinyl-epsilon-caprolactam, 99%, Thermo Scientific Chemicals
N-Vinyl-epsilon-caprolactam, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Vinyl-epsilon-caprolactam, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number B21311.18
also known as B21311-18
Price (USD)/ Each
89.65
Online exclusive
99.00 
Save 9.35 (9%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
89.65
Online exclusive
99.00 
Save 9.35 (9%)
Add to cart
N-Vinyl-epsilon-caprolactam, 99%, Thermo Scientific Chemicals
Catalog numberB21311.18
Price (USD)/ Each
89.65
Online exclusive
99.00 
Save 9.35 (9%)
-
Add to cart
Chemical Identifiers
CAS2235-00-9
IUPAC Name1-ethenylazepan-2-one
Molecular FormulaC8H13NO
InChI KeyJWYVGKFDLWWQJX-UHFFFAOYSA-N
SMILESC=CN1CCCCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥98.5%
Melting Point (clear melt)31.0-37.0?C
N-Vinyl-epsilon-caprolactam is used as reacive diluent in a number of our screen and is used in a wide range of UV curing screen printing inks. N-Vinyl-epsilon-caprolactam is used in UV-coatings and -adhesives as well as a building block for synthesis of paper coatings. Reactive diluent for in optical fiber coatings and rapid prototyping.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Vinyl-epsilon-caprolactam is used as reacive diluent in a number of our screen and is used in a wide range of UV curing screen printing inks. N-Vinyl-epsilon-caprolactam is used in UV-coatings and -adhesives as well as a building block for synthesis of paper coatings. Reactive diluent for in optical fiber coatings and rapid prototyping.

Solubility
Soluble in water (partly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only