1-Ethylguanidine sulfate may be used in chemical synthesis and thermal analysis studies. 1-Ethylguanidine sulfate is used as a reactant in the sunthesis of 3-Ethyl-6-methyl-isocytosines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Ethylguanidine sulfate may be used in chemical synthesis and thermal analysis studies. 1-Ethylguanidine sulfate is used as a reactant in the sunthesis of 3-Ethyl-6-methyl-isocytosines.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents,water, moisture.
RUO – Research Use Only
M. I. Fauth. Differential Thermal Analysis and Thermogravimetry of Some Salts of Guanidine and Related Compounds. Anal. Chem. 1960, 32 (6), 655-657.
Radu Custelceana,; Liliana Craciun. 3-Ethyl-6-methyl-isocytosines: Synthesis and Solid State Structural Analysis. Tetrahedron. 2000, 56 (29),5067-5075.