Thermo Scientific Chemicals

1,4-Diaminobutane, 98+%, Thermo Scientific Chemicals

Catalog number: B21316.14
25 g, Each
Thermo Scientific Chemicals

1,4-Diaminobutane, 98+%, Thermo Scientific Chemicals

Catalog number: B21316.14
25 g, Each
Quantity
Catalog number: B21316.14
also known as B21316-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-60-1
IUPAC Name
butane-1,4-diamine
Molecular Formula
C4H12N2
InChI Key
KIDHWZJUCRJVML-UHFFFAOYSA-N
SMILES
NCCCCN
Refractive Index
1.4540-1.4590 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid as melt
Identification (FTIR)
Conforms
Assay (GC)
≥98.0%

Description

1,4-Diaminobutane is used as a precursor in many biological systems and synthon for amido-ureas. It is involved in the synthesis of nylon 46 by reacting with adipic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diaminobutane is used as a precursor in many biological systems and synthon for amido-ureas. It is involved in the synthesis of nylon 46 by reacting with adipic acid.

Solubility
Soluble in ether and alcohol.

Notes
Incompatible with acids, strong oxidizing agents, aldehydes, acid anhydrides and acid chlorides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text