Cesium formate, 98%, Thermo Scientific Chemicals
Cesium formate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cesium formate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B21327.14
also known as B21327-14
Price (USD)/ Each
47.65
Online exclusive
52.40 
Save 4.75 (9%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
47.65
Online exclusive
52.40 
Save 4.75 (9%)
Add to cart
Cesium formate, 98%, Thermo Scientific Chemicals
Catalog numberB21327.14
Price (USD)/ Each
47.65
Online exclusive
52.40 
Save 4.75 (9%)
-
Add to cart
Chemical Identifiers
CAS3495-36-1
IUPAC Namecaesium(1+) formic acid
Molecular FormulaCH2CsO2
InChI KeyATZQZZAXOPPAAQ-UHFFFAOYSA-N
SMILES[Cs+].OC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or fused/lumpy solid or compacted powder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
CommentAs a bulk commodity material may contain a small amount of particulate matter from the manufacturing process
Water Content (Karl Fischer Titration)≤4.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only