Thermo Scientific Chemicals

4-Bromo-2-fluorobenzyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: B21331.14
25 g, Each
Thermo Scientific Chemicals

4-Bromo-2-fluorobenzyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: B21331.14
25 g, Each
Quantity
Catalog number: B21331.14
also known as B21331-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76283-09-5
IUPAC Name
4-bromo-1-(bromomethyl)-2-fluorobenzene
Molecular Formula
C7H5Br2F
InChI Key
XMHNLZXYPAULDF-UHFFFAOYSA-N
SMILES
FC1=CC(Br)=CC=C1CBr
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥97.5%
Melting Point (clear melt)
33.0-39.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text