Thermo Scientific Chemicals

Benzotrifluoride, 99%, Thermo Scientific Chemicals

Catalog number: B21340.30
250 g, Each
Thermo Scientific Chemicals

Benzotrifluoride, 99%, Thermo Scientific Chemicals

Catalog number: B21340.30
250 g, Each
Quantity
Catalog number: B21340.30
also known as B21340-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-08-8
IUPAC Name
(trifluoromethyl)benzene
Molecular Formula
C7H5F3
InChI Key
GETTZEONDQJALK-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=CC=C1
Assay (GC)
≥98.5%
Water Content (Karl Fischer Titration)
≤0.5%
Refractive Index
1.4130-1.4165 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid

Description

It is applied as an alternative solvent to dichloromethane, where it has a higher bp. It is used as an intermediate for dyes and pharmaceuticals, as a solvent and a dielectric fluid, a vulcanizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an alternative solvent to dichloromethane, where it has a higher bp. It is used as an intermediate for dyes and pharmaceuticals, as a solvent and a dielectric fluid, a vulcanizing agent.

Solubility
Soluble in water at 20°C0.45g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text