Thermo Scientific Chemicals

Methyl 2-hydroxy-3-methylbenzoate, 97%, Thermo Scientific Chemicals

Catalog number: B21341.09
10 g, Each
Thermo Scientific Chemicals

Methyl 2-hydroxy-3-methylbenzoate, 97%, Thermo Scientific Chemicals

Catalog number: B21341.09
10 g, Each
Quantity
Catalog number: B21341.09
also known as B21341-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23287-26-5
Appearance (Color)
Clear colorless to yellow to pale brown
Assay (GC)
≥96.0%
Refractive Index
1.5310-1.5360 @ 20°C
Form
Liquid

Description

Methyl 2-hydroxy-3-methylbenzoate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic intermediate. Some other applications include it is used to produce other chemicals. For example, it is used to produce 2-(2-Hydroxy-3-methyl-phenyl)-propan-2-ol. The reaction occurs with reagent Tetrahydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-hydroxy-3-methylbenzoate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic intermediate. Some other applications include it is used to produce other chemicals. For example, it is used to produce 2-(2-Hydroxy-3-methyl-phenyl)-propan-2-ol. The reaction occurs with reagent Tetrahydrofuran.

Solubility
Soluble in ethanol, fixed Oils, Propylene Glycol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Hygroscopic. Stable under normal temperatures and pressures.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text