3-Chloropropyl phenyl sulfide, 97%, Thermo Scientific Chemicals
3-Chloropropyl phenyl sulfide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chloropropyl phenyl sulfide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B21345.14
also known as B21345-14
Price (USD)/ Each
286.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
286.00
Add to cart
3-Chloropropyl phenyl sulfide, 97%, Thermo Scientific Chemicals
Catalog numberB21345.14
Price (USD)/ Each
286.00
-
Add to cart
Chemical Identifiers
CAS4911-65-3
IUPAC Name[(3-chloropropyl)sulfanyl]benzene
Molecular FormulaC9H11ClS
InChI KeySBRLITLVSMPPFI-UHFFFAOYSA-N
SMILESClCCCSC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5700-1.5750 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only