Thermo Scientific Chemicals

4-Fluorophthalic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: B21352.06
5 g, Each
Thermo Scientific Chemicals

4-Fluorophthalic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: B21352.06
5 g, Each
Quantity
Catalog number: B21352.06
also known as B21352-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
319-03-9
Appearance (Color)
White to pale cream
Form
powder
Assay (GC)
> 97.5%

Description

4-Fluorophthalic anhydride is used in the preparation of 5-fluoro-2-pyridin-3-ylmethyl-isoindole-1,3-dione by reacting with C-pyridin-3-yl-methylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophthalic anhydride is used in the preparation of 5-fluoro-2-pyridin-3-ylmethyl-isoindole-1,3-dione by reacting with C-pyridin-3-yl-methylamine.

Solubility
Soluble in sodium hydroxide.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text