Thermo Scientific Chemicals

1,3,5-Triphenylbenzene, 99+%, Thermo Scientific Chemicals

Catalog number: B21353.0B
1000 g, Each
Thermo Scientific Chemicals

1,3,5-Triphenylbenzene, 99+%, Thermo Scientific Chemicals

Catalog number: B21353.0B
1000 g, Each
Quantity
Catalog number: B21353.0B
also known as B21353-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
612-71-5
IUPAC Name
3,5-diphenyl-1,1'-biphenyl
Molecular Formula
C24H18
InChI Key
SXWIAEOZZQADEY-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream to yellow to pale brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥99.0%
Melting Point (clear melt)
171.0-177.0?C

Description

1,3,5-Triphenylbenzene fluorophore as a selective Cu2+ sensor in aqueous media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Triphenylbenzene fluorophore as a selective Cu2+ sensor in aqueous media.

Solubility
Solubility in toluene is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text