Thermo Scientific Chemicals

Boron trifluoride-dimethanol complex, 50-52% w/w boron trifluoride, Thermo Scientific Chemicals

Catalog number: B21357.AU
1000 mL, Each
Thermo Scientific Chemicals

Boron trifluoride-dimethanol complex, 50-52% w/w boron trifluoride, Thermo Scientific Chemicals

Catalog number: B21357.AU
1000 mL, Each
Quantity
Catalog number: B21357.AU
also known as B21357-AU
Price (TWD)

Chemical Identifiers

CAS
2802-68-8
IUPAC Name
bis(methanol); trifluoroborane
Molecular Formula
C2H8BF3O2
InChI Key
MPRMFRXCVMCOMX-UHFFFAOYSA-N
SMILES
CO.CO.FB(F)F
Appearance (Color)
Clear, colourless
Form
fuming liquid
Assay (unspecified)
BF3 content50 - 53% w/w
Density @20°C
1.210 - 1.235 kg/L

Description

It finds it application as a reagent for esterification of acids and also for the selective cleavage of trityl ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application as a reagent for esterification of acids and also for the selective cleavage of trityl ethers.

Solubility
Hydrolyzes in water.

Notes
^=50% boron fluoride in methanol
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text