4-(2-Aminoethyl)morpholine is used as a ligand and reacts with nickel(II) nitrite to form trans-bis[4-(2-aminoethyl)morpholine]dinitronickel(II).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(2-Aminoethyl)morpholine is used as a ligand and reacts with nickel(II) nitrite to form trans-bis[4-(2-aminoethyl)morpholine]dinitronickel(II).
Solubility
Miscible with water.
Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides and acid chlorides.
RUO – Research Use Only
Sheng, G. H.; Cheng, X. S.; You, Z. L.; Zhu, H. L. Synthesis and Crystal Structure of a mu-Oxido-Bridged Dinuclear Iron(III) Complex Derived From 3, 5-Dichlorosalicylaldehyde and 4-(2-Aminoethyl)morpholine. Synth React. Inorg. M. 2015, 45 (3), 388-391.
Wang, Q.; Zhou, L.; Qiu, L.; Lu, D.; Wu, Y.; Zhang, X. B. An efficient ratiometric fluorescent probe for tracking dynamic changes in lysosomal pH. Analyst 2015, 140 (16), 5563-5569.