Thermo Scientific Chemicals

3,5-Difluorobenzoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: B21400.14
25 g, Each
Thermo Scientific Chemicals

3,5-Difluorobenzoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: B21400.14
25 g, Each
Quantity
Catalog number: B21400.14
also known as B21400-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
129714-97-2
IUPAC Name
3,5-difluorobenzoyl chloride
Molecular Formula
C7H3ClF2O
InChI Key
OYZWEOORLJBPMA-UHFFFAOYSA-N
SMILES
FC1=CC(=CC(F)=C1)C(Cl)=O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.5015-1.5055 @ 20?C

Description

3,5-Difluorobenzoyl chloride, is used as an pharmaceutical intermediate, organic intermediate and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluorobenzoyl chloride, is used as an pharmaceutical intermediate, organic intermediate and chemical synthesis.

Solubility
It reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong acids and bases. Moisture Sensitive
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text