Thermo Scientific Chemicals

Diethyl terephthalate, 95%, Thermo Scientific Chemicals

Catalog number: B21401.18
50 g, Each
Thermo Scientific Chemicals

Diethyl terephthalate, 95%, Thermo Scientific Chemicals

Catalog number: B21401.18
50 g, Each
Quantity
Catalog number: B21401.18
also known as B21401-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
636-09-9
IUPAC Name
1,4-diethyl benzene-1,4-dicarboxylate
Molecular Formula
C12H14O4
InChI Key
ONIHPYYWNBVMID-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=C(C=C1)C(=O)OCC
Assay (GC)
≥94.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
39.0-51.0?C
Appearance (Color)
White to pale cream to cream to yellow or pink
Form
Crystals or powder or crystalline powder or fused solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text